Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098525.2 | NP_001091995.1 | 2 | 298 | ||
NM_001286686.1 | NP_001273615.1 | 2 | 239 | |||
NM_001286687.1 | NP_001273616.1 | 2 | 298 | |||
NM_018204.4 | NP_060674.3 | 2 | 298 | |||
XM_005266344.4 | XP_005266401.1 | 2 | 206 | |||
XM_011535043.2 | XP_011533345.1 | 2 | 229 | |||
GenBank mRNA | AK022982.1 | - | 2 | 197 | ||
AK299083.1 | - | 2 | 239 | |||
AY062261.1 | - | 2 | 219 | |||
AY062262.1 | - | 2 | 219 | |||
BC105806.1 | - | 2 | 177 | |||
BC130296.1 | - | 2 | 146 | 40146327 | ||
BC136332.1 | - | 2 | 146 | 9020319 | ||
BM541375.1 | - | 2 | 303 | |||
EF560732.1 | - | 2 | 179 | |||
Y15758.1 | - | 2 | 191 |