Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080449.2 | NP_001073918.2 | 10 | 1714 | ||
NR_102264.1 | 11 | 1694 | ||||
XM_006717680.3 | XP_006717743.1 | 11 | 2094 | |||
XM_011539417.1 | XP_011537719.1 | 4 | 638 | |||
XM_017015799.1 | XP_016871288.1 | 7 | 1103 | |||
GenBank mRNA | AK308333.1 | - | 10 | 1714 | ||
AK308358.1 | - | 8 | 1355 | |||
BC063664.1 | - | 10 | 1627 | |||
BC111740.1 | - | 11 | 1694 | 40038149 | ||
D42046.1 | - | 10 | 1656 |