Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other METTL23 SIRNA Assays ›
Gene Symbol:
METTL23
Gene Name:
methyltransferase like 23
Gene Aliases:
C17orf95, MRT44
Chromosome Location:
Chr.17: 76725874 - 76733881 on Build GRCh38
UniGene ID:
Hs.74655
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
263866
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: METTL23
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080510.4
NP_001073979.3
3, 4
694
NM_001206983.2
NP_001193912.1
3, 4
729
NM_001206984.2
NP_001193913.1
3, 4
395
NM_001206985.2
NP_001193914.1
2, 3
589
NM_001206986.2
NP_001193915.1
2, 3
624
NM_001206987.2
NP_001193916.1
2, 3
768
NM_001302703.1
NP_001289632.1
4, 5
639
NM_001302704.1
NP_001289633.1
3, 4
499
NM_001302705.1
NP_001289634.1
3, 4
378
XM_006721674.3
XP_006721737.1
3, 4
757
XM_006721675.1
XP_006721738.1
3, 4
621
XM_006721676.3
XP_006721739.1
3, 4
656
XM_006721678.3
XP_006721741.1
3, 4
742
XM_006721679.3
XP_006721742.1
2, 3
387
XM_006721680.2
XP_006721743.1
2, 3
296
XM_011524282.1
XP_011522584.1
3, 4
485
XM_017024145.1
XP_016879634.1
4, 5
589
XM_017024146.1
XP_016879635.1
3, 4
499
GenBank mRNA
AL522160.3
-
3, 4
679
BC033998.2
-
2, 3
565
BC045819.1
-
3, 4
656
BE567652.1
-
3, 4
350
BF575183.1
-
3, 4
396
BI225157.1
-
2, 3
588
BI819093.1
-
3, 4
689
BI829268.1
-
2, 3
550
BU176249.1
-
3, 4
381
BU182031.1
-
2, 3
601
BX356072.2
-
3, 4
478
BX396749.2
-
4, 5
626
BX403283.2
-
3, 4
505
JQ313903.1
-
2, 3
324
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
methylation
methyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0