Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLPH3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018178.5 | NP_060648.2 | 2 | 351 | ||
XM_006711428.1 | XP_006711491.1 | 2 | 278 | |||
GenBank mRNA | AJ296153.1 | - | 2 | 179 | ||
AK300635.1 | - | 2 | 309 | |||
AK315594.1 | - | 2 | 318 | |||
BC013870.2 | - | 1 | 150 | 3837466 | ||
BI550986.1 | - | 322 | ||||
DA513375.1 | - | 2 | 351 |