Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCDC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099434.1 | NP_001092904.1 | 4 | 491 | ||
XM_017000414.1 | XP_016855903.1 | 2 | 482 | |||
GenBank mRNA | BC041458.1 | - | 3 | 721 | ||
BC128073.1 | - | 4 | 491 | 40109068 |