Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033931.1 | 3 | 1348 | |||
GenBank mRNA | AK055518.1 | - | 3 | 1349 | ||
BC121142.1 | - | 2 | 1113 | |||
BC121143.1 | - | 2 | 1109 |