Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085429.1 | NP_001078898.1 | 3 | 509 | ||
XM_005250167.3 | XP_005250224.1 | 3 | 1013 | |||
GenBank mRNA | AK055841.1 | - | 3 | 509 | ||
BC131697.1 | - | 3 | 552 | 40120595 |