Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM65C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290268.1 | NP_001277197.1 | 22 | 3281 | ||
NM_080829.3 | NP_543019.2 | 22 | 3214 | |||
NR_110890.1 | 22 | 3440 | ||||
XM_005260294.3 | XP_005260351.1 | 23 | 3278 | |||
XM_006723713.3 | XP_006723776.1 | 22 | 3326 | |||
XM_011528578.2 | XP_011526880.1 | 23 | 3306 | |||
XM_011528579.2 | XP_011526881.1 | 23 | 3444 | |||
XM_011528580.2 | XP_011526882.1 | 23 | 3498 | |||
XM_011528581.2 | XP_011526883.1 | 23 | 3482 | |||
XM_011528585.2 | XP_011526887.1 | 12 | 2629 | |||
XM_017027681.1 | XP_016883170.1 | 14 | 2274 | |||
GenBank mRNA | AK299337.1 | - | 22 | 3130 | ||
CA389799.1 | - | 3 | 266 | |||
CD558804.1 | - | 3 | 308 |