Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039360.2 | NP_001034449.1 | 2 | 1260 | ||
NM_001318841.1 | NP_001305770.1 | 4 | 1441 | |||
XM_005258229.4 | XP_005258286.1 | 3 | 1185 | |||
XM_011525861.2 | XP_011524163.1 | 6 | 1665 | |||
XM_011525863.2 | XP_011524165.1 | 5 | 1782 | |||
XM_011525864.2 | XP_011524166.1 | 5 | 1537 | |||
XM_011525865.2 | XP_011524167.1 | 5 | 1778 | |||
XM_011525866.2 | XP_011524168.1 | 4 | 1523 | |||
XM_011525867.2 | XP_011524169.1 | 4 | 1312 | |||
XM_011525869.2 | XP_011524171.1 | 5 | 1787 | |||
XM_011525870.2 | XP_011524172.1 | 4 | 1483 | |||
XM_011525871.2 | XP_011524173.1 | 2 | 1998 | |||
XM_017025605.1 | XP_016881094.1 | 6 | 3460 | |||
XM_017025606.1 | XP_016881095.1 | 6 | 1914 | |||
XM_017025607.1 | XP_016881096.1 | 6 | 1669 | |||
XM_017025608.1 | XP_016881097.1 | 5 | 3337 | |||
XM_017025609.1 | XP_016881098.1 | 4 | 1654 | |||
GenBank mRNA | AM988836.1 | - | 2 | 1248 | ||
AM988837.1 | - | 2 | 1393 | |||
AM988838.1 | - | 3 | 1443 | |||
AM988839.1 | - | 4 | 1441 | |||
EF120357.1 | - | 2 | 1312 |