Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TBC1D12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015188.1 | NP_056003.1 | 13 | 3607 | ||
XM_006717732.3 | XP_006717795.2 | 12 | 3526 | |||
XM_006717733.2 | XP_006717796.1 | 14 | 2567 | |||
XM_011539558.2 | XP_011537860.1 | 13 | 2618 | |||
XM_017015996.1 | XP_016871485.1 | 11 | 3382 | |||
XM_017015998.1 | XP_016871487.1 | 12 | 2394 | |||
XM_017015999.1 | XP_016871488.1 | 12 | 2610 | |||
GenBank mRNA | AJ404330.1 | - | 1 | 158 | ||
BC064323.1 | - | 4 | 1625 |