Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304333.2 | NP_001291262.1 | 8 | 4905 | ||
NM_015297.2 | NP_056112.1 | 8 | 4901 | |||
XM_011517827.1 | XP_011516129.1 | 9 | 4893 | |||
XM_011517828.2 | XP_011516130.1 | 8 | 4997 | |||
XM_011517829.1 | XP_011516131.1 | 8 | 5155 | |||
XM_017014553.1 | XP_016870042.1 | 10 | 5187 | |||
XM_017014554.1 | XP_016870043.1 | 10 | 4967 | |||
XM_017014555.1 | XP_016870044.1 | 10 | 4939 | |||
XM_017014556.1 | XP_016870045.1 | 10 | 5471 | |||
GenBank mRNA | AB028968.1 | - | 8 | 4767 | ||
AL137621.1 | - | 1 | 432 |