Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2SURP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080415.1 | NP_001073884.1 | 16 | 1579 | ||
NM_001320219.1 | NP_001307148.1 | 16 | 1576 | |||
NM_001320220.1 | NP_001307149.1 | 15 | 1535 | |||
NM_001320222.1 | NP_001307151.1 | 17 | 1644 | |||
XM_005247248.4 | XP_005247305.1 | 16 | 1585 | |||
XM_017006037.1 | XP_016861526.1 | 12 | 1187 | |||
XM_017006038.1 | XP_016861527.1 | 15 | 1471 | |||
XM_017006039.1 | XP_016861528.1 | 9 | 914 | |||
GenBank mRNA | AB002330.1 | - | 16 | 1478 | ||
AK057679.1 | - | 3 | 1573 | |||
AK074327.1 | - | 9 | 843 | |||
BC006474.1 | - | 16 | 1475 | |||
BC111692.1 | - | 15 | 1906 | 40028600 |