Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2SURP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080415.1 | NP_001073884.1 | 28 | 6284 | ||
NM_001320219.1 | NP_001307148.1 | 28 | 6281 | |||
NM_001320220.1 | NP_001307149.1 | 27 | 6240 | |||
NM_001320222.1 | NP_001307151.1 | 29 | 6349 | |||
XM_005247248.4 | XP_005247305.1 | 27 | 6113 | |||
XM_017006037.1 | XP_016861526.1 | 24 | 5892 | |||
XM_017006038.1 | XP_016861527.1 | 27 | 6176 | |||
XM_017006039.1 | XP_016861528.1 | 21 | 5619 | |||
GenBank mRNA | AB002330.1 | - | 28 | 6185 | ||
BC045744.1 | - | 1 | 1770 |