Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM222B SIRNA Assays ›
Gene Symbol:
FAM222B
Gene Name:
family with sequence similarity 222 member B
Gene Aliases:
C17orf63
Chromosome Location:
Chr.17: 28755978 - 28855036 on Build GRCh38
UniGene ID:
Hs.564533
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
26516
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM222B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077498.2
NP_001070966.1
3
2441
NM_001288631.1
NP_001275560.1
4
2612
NM_001288632.1
NP_001275561.1
5
2523
NM_001288633.1
NP_001275562.1
4
2517
NM_001288634.1
NP_001275563.1
3
2510
NM_001288635.1
NP_001275564.1
4
2489
NM_001288636.1
NP_001275565.1
6
2513
NM_001288637.1
NP_001275566.1
5
2479
NM_001288638.1
NP_001275567.1
5
2411
NM_001288639.1
NP_001275568.1
4
2363
NM_001288640.1
NP_001275569.1
3
2329
NM_018182.3
NP_060652.2
4
2475
XM_011524986.2
XP_011523288.1
3
2406
XM_011524987.1
XP_011523289.1
4
2241
XM_011524988.2
XP_011523290.1
3
2457
XM_017024831.1
XP_016880320.1
4
2565
XM_017024832.1
XP_016880321.1
3
2830
XM_017024833.1
XP_016880322.1
3
2370
XM_017024834.1
XP_016880323.1
3
2568
XM_017024835.1
XP_016880324.1
4
2767
GenBank mRNA
AK000147.1
-
1
1585
AK001562.1
-
4
2426
AK025980.1
-
1
1444
AK057665.1
-
3
2425
AK095569.1
-
3
2510
BC021228.2
-
3
2325
4040385
BX647766.1
-
3
2260
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0