Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A25-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033374.1 | 2 | 848 | |||
GenBank mRNA | AK124723.1 | - | 2 | 846 | ||
AL833509.1 | - | 2 | 807 |