Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPEPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018226.4 | NP_060696.4 | 11 | 2679 | ||
XM_005247036.4 | XP_005247093.1 | 11 | 2707 | |||
GenBank mRNA | AF300795.1 | - | 11 | 2385 | ||
AK001668.1 | - | 7 | 1643 | |||
AK130185.1 | - | 4 | 1247 | |||
AK130805.1 | - | 4 | 3613 | |||
AL512754.1 | - | 4 | 1264 | |||
BC017301.2 | - | 8 | 1831 | |||
BC067258.1 | - | 11 | 2254 | 6183046 | ||
BC082975.1 | - | 11 | 2579 | 6379537 | ||
BC099906.1 | - | 9 | 2792 | 5311883 |