Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304530.1 | NP_001291459.1 | 5, 6 | 407 | ||
NM_001304532.1 | NP_001291461.1 | 5, 6 | 407 | |||
NM_018246.2 | NP_060716.2 | 6, 7 | 459 | |||
XM_011544570.2 | XP_011542872.1 | 5, 6 | 406 | |||
XM_011544571.2 | XP_011542873.1 | 4, 5 | 285 | |||
GenBank mRNA | AK001715.1 | - | 5, 6 | 407 | ||
AK027900.1 | - | 6, 7 | 411 | |||
AK292875.1 | - | 5, 6 | 394 | |||
BC006239.2 | - | 7, 8 | 634 | |||
BC032588.1 | - | 5, 6 | 396 | 5502227 | ||
BC049197.1 | - | 6, 7 | 413 | 5534448 | ||
BC054039.1 | - | 6, 7 | 399 | |||
BC065295.1 | - | 6, 7 | 400 | 6139413 | ||
BG718771.1 | - | 5, 6 | 399 | |||
DA218015.1 | - | 6, 7 | 459 |