Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145474.3 | NP_001138946.1 | 2 | 686 | ||
NM_001300863.1 | NP_001287792.1 | 2 | 607 | |||
NM_001300864.1 | NP_001287793.1 | 2 | 535 | |||
XM_011541421.2 | XP_011539723.1 | 2 | 682 | |||
GenBank mRNA | BC057818.1 | - | 2 | 665 | ||
BC067853.1 | - | 2 | 691 | |||
BC126318.1 | - | 2 | 641 | |||
BC144026.1 | - | 2 | 562 | 9052546 | ||
BC144027.1 | - | 2 | 490 | 9052547 | ||
FJ211596.1 | - | 2 | 584 |