Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C10orf67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011519443.1 | XP_011517745.1 | 12 | 1107 | ||
XM_017016029.1 | XP_016871518.1 | 16 | 2248 | |||
XM_017016030.1 | XP_016871519.1 | 15 | 1650 | |||
XM_017016031.1 | XP_016871520.1 | 15 | 1661 |