Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039752.3 | NP_001034841.3 | 5 | 1082 | ||
NR_134874.1 | 6 | 837 | ||||
XM_011545016.2 | XP_011543318.1 | 7 | 2087 | |||
XM_017017699.1 | XP_016873188.1 | 6 | 884 | |||
XM_017017700.1 | XP_016873189.1 | 5 | 1320 | |||
XM_017017701.1 | XP_016873190.1 | 7 | 2087 | |||
XM_017017702.1 | XP_016873191.1 | 7 | 2087 | |||
XM_017017703.1 | XP_016873192.1 | 4 | 428 | |||
XM_017017704.1 | XP_016873193.1 | 5 | 1319 | |||
GenBank mRNA | AB075876.1 | - | 6 | 837 | ||
AK296299.1 | - | 7 | 789 | |||
AK298430.1 | - | 6 | 884 | |||
BC171744.1 | - | 3 | 417 | |||
DR003989.1 | - | 5 | 616 |