Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018328.4 | NP_060798.2 | 9 | 3084 | ||
XM_011511470.2 | XP_011509772.1 | 9 | 3316 | |||
XM_011511472.2 | XP_011509774.1 | 7 | 2896 | |||
XM_017004482.1 | XP_016859971.1 | 5 | 2766 | |||
XM_017004483.1 | XP_016859972.1 | 5 | 2766 | |||
XM_017004484.1 | XP_016859973.1 | 5 | 2766 | |||
XM_017004485.1 | XP_016859974.1 | 5 | 2766 | |||
GenBank mRNA | AB040894.1 | - | 4 | 2377 | ||
AK304424.1 | - | 4 | 2399 | |||
BC014534.1 | - | 1 | 533 | |||
BC150264.1 | - | 4 | 2377 | 9007212 | ||
EF542797.1 | - | 9 | 3075 |