Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCF2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011537482.1 | XP_011535784.1 | 2 | 186 | ||
XM_011537483.2 | XP_011535785.1 | 2 | 306 |