Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RHOT1 SIRNA Assays ›
Gene Symbol:
RHOT1
ANKRD20A9P
Gene Name:
ras homolog family member T1
ankyrin repeat domain 20 family member A9, pseudogene
Gene Aliases:
ARHT1, MIRO-1, MIRO1
-
Chromosome Location:
Chr.17: 32142454 - 32253374 on Build GRCh38
Chromosome Location:
Chr.13: 18834403 - 18871969 on Build GRCh38
UniGene ID:
Hs.655325
Hs.679496
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
26710
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RHOT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001033566.2
NP_001028738.1
5, 6
523
NM_001033567.2
NP_001028739.2
6, 7
647
NM_001033568.2
NP_001028740.1
5, 6
523
NM_001288754.1
NP_001275683.1
5, 6
523
NM_001288755.1
NP_001275684.1
5, 6
519
NM_001288758.1
NP_001275687.1
6, 7
647
NM_018307.4
NP_060777.3
5, 6
523
NR_110083.1
5, 6
523
XM_011524969.2
XP_011523271.1
4, 5
247
XM_011524971.2
XP_011523273.2
5, 6
319
XM_011524973.2
XP_011523275.2
5, 6
319
XM_011524975.2
XP_011523277.1
6, 7
776
XM_011524976.2
XP_011523278.2
5, 6
319
XM_017024819.1
XP_016880308.1
6, 7
551
XM_017024820.1
XP_016880309.1
4, 5
247
XM_017024821.1
XP_016880310.1
5, 6
319
XM_017024822.1
XP_016880311.1
5, 6
548
XM_017024823.1
XP_016880312.1
5, 6
319
XM_017024824.1
XP_016880313.1
6, 7
654
XM_017024825.1
XP_016880314.1
6, 7
746
GenBank mRNA
AJ496730.1
-
5, 6
511
AJ517412.1
-
5, 6
284
AK294407.1
-
5, 6
297
AL136929.1
-
5, 6
347
AY094972.1
-
5, 6
400
BC015698.2
-
1
17
BC041114.1
-
5, 6
457
BC051818.1
-
4, 5
234
BC060781.1
-
5, 6
523
4823115
BC068463.1
-
5, 6
411
BC125104.1
-
5, 6
352
40118340
BC125105.1
-
5, 6
352
40118341
CN352790.1
-
5, 6
480
Gene Symbol: ANKRD20A9P
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_027995.1
1
34154
Back To Top
More Information
Panther Classification:
Molecular Function -
small GTPase
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
cellular homeostasis
mitochondrion transport along microtubule
regulation of small GTPase mediated signal transduction
mitochondrial outer membrane permeabilization
molecular_function
GTPase activity
calcium ion binding
protein binding
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0