Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STEAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008410.1 | NP_001008410.1 | 5 | 3292 | ||
NM_018234.2 | NP_060704.2 | 6 | 3360 | |||
NM_138637.2 | NP_619543.2 | 6 | 3524 | |||
NM_182915.2 | NP_878919.2 | 6 | 3707 | |||
XM_006712614.3 | XP_006712677.1 | 6 | 3434 | |||
XM_006712615.1 | XP_006712678.1 | 5 | 3304 | |||
XM_011511403.1 | XP_011509705.1 | 7 | 3718 | |||
GenBank mRNA | AF423424.1 | - | 6 | 3337 | ||
AK001691.1 | - | 6 | 3337 | |||
AK024163.1 | - | 4 | 2249 | |||
AL833624.1 | - | 6 | 3707 | |||
AY214461.1 | - | 6 | 3337 | |||
BC095421.1 | - | 5 | 3247 | 30336214 | ||
BX538047.1 | - | 6 | 3676 |