Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM184C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018241.2 | NP_060711.2 | 10 | 1672 | ||
GenBank mRNA | AF305823.1 | - | 3 | 348 | ||
AK000169.1 | - | 7 | 825 | |||
AK001708.1 | - | 10 | 1615 | |||
BC008120.1 | - | 2 | 204 | |||
BM712423.1 | - | 4 | 392 |