Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OACYLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024021.2 | 4 | 398 | |||
GenBank mRNA | DA088392.1 | - | 2 | 44 | ||
DV080257.1 | - | 4 | 338 |