Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199198.2 | NP_001186127.1 | 18, 19 | 2228 | ||
NM_018309.4 | NP_060779.2 | 17, 18 | 2183 | |||
XM_011512974.2 | XP_011511276.1 | 18, 19 | 2091 | |||
XM_017006840.1 | XP_016862329.1 | 17, 18 | 1889 | |||
XM_017006841.1 | XP_016862330.1 | 18, 19 | 2059 | |||
GenBank mRNA | AB449910.1 | - | 17, 18 | 1981 | ||
AK001750.1 | - | 14, 15 | 1669 | |||
AK001908.1 | - | 17, 18 | 1988 | |||
AK092926.1 | - | 3, 4 | 664 | |||
AL713684.1 | - | 10, 11 | 1109 | |||
AY740521.1 | - | 7, 8 | 889 | |||
BC010221.1 | - | 8, 9 | 926 | |||
BC020955.1 | - | 18, 19 | 2022 | 3847561 | ||
BX648032.1 | - | 17, 18 | 1975 |