Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300926.1 | NP_001287855.1 | 7 | 2089 | ||
NM_018320.4 | NP_060790.2 | 9 | 2251 | |||
NR_024147.1 | 8 | 2213 | ||||
NR_024148.1 | 9 | 2351 | ||||
XM_006718629.1 | XP_006718692.1 | 8 | 2164 | |||
XM_006718630.1 | XP_006718693.1 | 10 | 2064 | |||
XM_006718632.1 | XP_006718695.1 | 7 | 2009 | |||
XM_011545150.1 | XP_011543452.1 | 10 | 2096 | |||
GenBank mRNA | AK001961.1 | - | 4 | 1413 | ||
AK023139.1 | - | 9 | 1915 | |||
AK094508.1 | - | 9 | 2015 | |||
AL833638.1 | - | 7 | 1762 | |||
BC093084.1 | - | 8 | 1851 |