Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018346.2 | NP_060816.1 | 9 | 2136 | ||
NR_130911.1 | 5 | 1775 | ||||
GenBank mRNA | AK002026.1 | - | 9 | 2107 | ||
AK022105.1 | - | 9 | 2076 | |||
AK024628.1 | - | 4 | 1116 | |||
BC005854.2 | - | 8 | 1888 | |||
BC050538.1 | - | 9 | 2067 | 5214885 | ||
CF130211.1 | - | 1 | 244 |