Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF415 SIRNA Assays ›
Gene Symbol:
ZNF415
Gene Name:
zinc finger protein 415
Gene Aliases:
ZfLp
Chromosome Location:
Chr.19: 53107879 - 53133077 on Build GRCh38
UniGene ID:
Hs.147765
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
26839
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF415
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136038.2
NP_001129510.2
4
534
NM_001164309.1
NP_001157781.1
4
407
NM_018355.3
NP_060825.2
4
302
NR_028343.1
7
663
XM_006723267.1
XP_006723330.1
6
620
XM_006723268.1
XP_006723331.1
4
368
XM_006723272.2
XP_006723335.1
5
400
XM_006723273.2
XP_006723336.1
5
484
XM_006723274.2
XP_006723337.1
5
561
XM_006723277.2
XP_006723340.1
5
495
XM_006723279.1
XP_006723342.1
5
654
XM_006723280.2
XP_006723343.1
2
110
XM_006723281.2
XP_006723344.1
3
136
XM_006723282.1
XP_006723345.1
3
445
XM_011527100.1
XP_011525402.1
5
644
XM_011527101.2
XP_011525403.1
5
619
XM_011527103.1
XP_011525405.1
5
439
XM_011527104.1
XP_011525406.1
5
733
XM_017026952.1
XP_016882441.1
5
447
XM_017026953.1
XP_016882442.1
6
767
XM_017026954.1
XP_016882443.1
6
644
XM_017026955.1
XP_016882444.1
6
595
XM_017026956.1
XP_016882445.1
7
745
XM_017026957.1
XP_016882446.1
6
483
XM_017026958.1
XP_016882447.1
7
799
XM_017026959.1
XP_016882448.1
6
670
XM_017026960.1
XP_016882449.1
5
385
XM_017026961.1
XP_016882450.1
6
831
XM_017026962.1
XP_016882451.1
6
676
XM_017026963.1
XP_016882452.1
5
572
XM_017026964.1
XP_016882453.1
5
522
XM_017026965.1
XP_016882454.1
4
536
XM_017026966.1
XP_016882455.1
5
664
XM_017026967.1
XP_016882456.1
4
462
XM_017026968.1
XP_016882457.1
6
760
XM_017026969.1
XP_016882458.1
2
181
XM_017026970.1
XP_016882459.1
6
637
XM_017026971.1
XP_016882460.1
6
856
XM_017026972.1
XP_016882461.1
4
438
XM_017026973.1
XP_016882462.1
5
630
XM_017026974.1
XP_016882463.1
6
709
GenBank mRNA
AK002053.1
-
4
274
AK095504.1
-
7
635
AL534796.3
-
5
484
AY283600.1
-
4
274
BC063880.1
-
5
485
30388597
BC069668.1
-
5
484
7262632
DA075573.1
-
4
389
DA367299.1
-
3
274
DA518713.1
-
4
504
DB285275.1
-
4
408
DQ925695.1
-
3
136
DQ925696.1
-
5
473
DQ925697.1
-
6
483
DQ925698.1
-
5
585
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0