Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTATIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098520.1 | NP_001091990.1 | 2 | 356 | ||
NM_001098521.1 | NP_001091991.1 | 2 | 284 | |||
NM_001098522.1 | NP_001091992.1 | 1 | 566 | |||
NM_001098523.1 | NP_001091993.1 | 1 | 282 | |||
NM_006410.4 | NP_006401.3 | 2 | 251 | |||
GenBank mRNA | AF039103.1 | - | 1 | 197 | ||
AF092095.1 | - | 1 | 283 | |||
AK222969.1 | - | 1 | 240 | |||
AK223010.1 | - | 1 | 274 | |||
AK292092.1 | - | 2 | 260 | |||
AW963626.1 | - | 1 | 201 | |||
BC002439.2 | - | 2 | 297 | 3346984 | ||
BC015358.2 | - | 1 | 262 | 3455757 | ||
BF681178.1 | - | 250 | ||||
BP382529.1 | - | 2 | 356 | |||
CR456821.1 | - | 1 | 185 | |||
DB450790.1 | - | 2 | 283 | |||
U69161.1 | - | 1 | 283 |