Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018370.2 | NP_060840.2 | 7 | 1157 | ||
XM_005269004.2 | XP_005269061.1 | 6 | 949 | |||
XM_005269005.2 | XP_005269062.1 | 5 | 920 | |||
XM_017019578.1 | XP_016875067.1 | 8 | 1279 | |||
GenBank mRNA | AK002121.1 | - | 5 | 463 | ||
AK094923.1 | - | 7 | 787 | |||
AK297785.1 | - | 4 | 471 | |||
BC013773.1 | - | 5 | 455 | |||
BC018435.1 | - | 7 | 1133 | 4474297 |