Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNJ2BP-COX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202547.2 | NP_001189476.1 | 2 | 315 | ||
NM_001202548.2 | NP_001189477.1 | 2 | 315 | |||
NM_001202549.2 | NP_001189478.1 | 2 | 315 | |||
GenBank mRNA | FY210787.1 | - | 2 | 209 | ||
FY213292.1 | - | 2 | 209 | |||
FY213836.1 | - | 2 | 209 |
Gene Symbol: SYNJ2BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018373.2 | NP_060843.2 | 2 | 315 | ||
GenBank mRNA | AK002133.1 | - | 2 | 270 | ||
AY534623.1 | - | 2 | 284 | |||
BC007704.2 | - | 2 | 217 | 3952366 | ||
BP195583.1 | - | 2 | 315 |