Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPR1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108075.1 | 1 | 267 | |||
GenBank mRNA | AL832729.1 | - | 1 | 279 | ||
BI523552.1 | - | 1 | 295 |