Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ST6GALNAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289107.1 | NP_001276036.1 | 1 | 316 | ||
NM_018414.4 | NP_060884.1 | 1 | 316 | |||
NR_110309.1 | 1 | 316 | ||||
XM_011524995.2 | XP_011523297.1 | 1 | 236 | |||
XM_011524996.2 | XP_011523298.1 | 1 | 236 | |||
XM_017024842.1 | XP_016880331.1 | 1 | 236 | |||
XM_017024843.1 | XP_016880332.1 | 1 | 236 | |||
XM_017024844.1 | XP_016880333.1 | 1 | 199 | |||
GenBank mRNA | AK310906.1 | - | 1 | 252 | ||
AY096001.1 | - | 1 | 116 | |||
AY358918.1 | - | 1 | 261 | |||
BC022462.1 | - | 1 | 171 | 4816713 | ||
DA594489.1 | - | 1 | 316 | |||
Y11339.2 | - | 1 | 176 |