Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267571.1 | NP_001254500.1 | 7 | 1640 | ||
NM_001267572.1 | NP_001254501.1 | 1 | 419 | |||
NM_018421.3 | NP_060891.3 | 7 | 1640 | |||
NR_051978.1 | 6 | 1498 | ||||
XM_011518843.1 | XP_011517145.1 | 2 | 1522 | |||
XM_017014899.1 | XP_016870388.1 | 7 | 1070 | |||
GenBank mRNA | AB449882.1 | - | 7 | 1460 | ||
AF318370.1 | - | 7 | 1499 | |||
AK026105.1 | - | 6 | 988 | |||
AK124772.1 | - | 6 | 2189 | |||
AK131290.1 | - | 7 | 1518 | |||
AK298575.1 | - | 8 | 1290 | |||
AL353935.1 | - | 2 | 1701 | |||
AY026527.1 | - | 7 | 1552 | |||
BC028918.1 | - | 1 | 419 | 4931447 | ||
BC071978.1 | - | 3 | 527 | 5443205 |