Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308277.1 | NP_001295206.1 | 3, 4 | 512 | ||
NM_018381.3 | NP_060851.2 | 3, 4 | 512 | |||
GenBank mRNA | AK001098.1 | - | 218 | |||
AK002148.1 | - | 3, 4 | 298 | |||
AL137694.1 | - | 3, 4 | 214 | |||
BC035817.2 | - | 3, 4 | 304 | 5554801 |