Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCKAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013436.4 | NP_038464.1 | 3, 4 | 1079 | ||
NM_205842.2 | NP_995314.1 | 4, 5 | 1097 | |||
XM_006712200.3 | XP_006712263.1 | 4, 5 | 583 | |||
XM_006712201.3 | XP_006712264.1 | 3, 4 | 567 | |||
GenBank mRNA | AB011159.1 | - | 4, 5 | 1097 | ||
AB014509.1 | - | 3, 4 | 339 | |||
AK292914.1 | - | 3, 4 | 569 | |||
BC015025.1 | - | 3, 4 | 591 | 3907646 | ||
BT007033.1 | - | 3, 4 | 320 |