Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSGALNACT1 SIRNA Assays ›
Gene Symbol:
CSGALNACT1
Gene Name:
chondroitin sulfate N-acetylgalactosaminyltransferase 1
Gene Aliases:
CSGalNAcT-1, ChGn, beta4GalNAcT
Chromosome Location:
Chr.8: 19404161 - 19757940 on Build GRCh38
UniGene ID:
Hs.613729
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
27045
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CSGALNACT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130518.1
NP_001123990.1
10
3602
NM_018371.4
NP_060841.5
10
3192
NR_024040.1
10
3271
XM_006716358.2
XP_006716421.1
9
3483
XM_006716359.2
XP_006716422.1
10
3852
XM_006716360.2
XP_006716423.1
11
4000
XM_006716361.2
XP_006716424.1
10
3747
XM_006716362.2
XP_006716425.1
9
3070
XM_006716363.1
XP_006716426.1
9
3242
XM_006716364.3
XP_006716427.1
8
2926
XM_011544578.1
XP_011542880.1
12
4119
XM_011544579.1
XP_011542881.1
11
3971
XM_011544580.1
XP_011542882.1
10
3849
XM_011544582.1
XP_011542884.1
10
3857
XM_011544583.1
XP_011542885.1
8
3036
XM_011544584.1
XP_011542886.1
8
2920
XM_011544585.2
XP_011542887.1
7
2003
XM_017013623.1
XP_016869112.1
13
4169
XM_017013624.1
XP_016869113.1
10
3808
XM_017013625.1
XP_016869114.1
8
3085
XM_017013626.1
XP_016869115.1
9
3015
XM_017013627.1
XP_016869116.1
10
5409
XM_017013628.1
XP_016869117.1
9
5269
XM_017013629.1
XP_016869118.1
10
3200
XM_017013630.1
XP_016869119.1
10
3113
XM_017013631.1
XP_016869120.1
9
3074
XM_017013632.1
XP_016869121.1
10
3183
XM_017013633.1
XP_016869122.1
10
3170
XM_017013634.1
XP_016869123.1
10
3167
XM_017013635.1
XP_016869124.1
10
3175
XM_017013636.1
XP_016869125.1
11
3318
XM_017013637.1
XP_016869126.1
10
3175
XM_017013638.1
XP_016869127.1
11
3360
XM_017013639.1
XP_016869128.1
9
3008
GenBank mRNA
AB081516.1
-
8
2880
AK002124.1
-
1
951
AK002126.1
-
7
2129
AL157483.1
-
3
1399
AY358441.1
-
10
3192
BC060772.1
-
10
3284
30340553
BX104931.1
-
1
227
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
endochondral ossification
nervous system development
cell recognition
cell proliferation
anatomical structure morphogenesis
heparan sulfate proteoglycan biosynthetic process, polysaccharide chain biosynthetic process
UDP-N-acetylgalactosamine metabolic process
proteoglycan biosynthetic process
extracellular matrix organization
chondroitin sulfate biosynthetic process
heparin biosynthetic process
UDP-glucuronate metabolic process
chondroitin sulfate proteoglycan biosynthetic process
dermatan sulfate proteoglycan biosynthetic process
chondroitin sulfate proteoglycan biosynthetic process, polysaccharide chain biosynthetic process
cartilage development
acetylgalactosaminyltransferase activity
peptidoglycan glycosyltransferase activity
glucuronosyltransferase activity
metal ion binding
glucuronylgalactosylproteoglycan 4-beta-N-acetylgalactosaminyltransferase activity
glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0