Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018378.2 | NP_060848.2 | 3 | 1576 | ||
GenBank mRNA | AK002140.1 | - | 3 | 1577 | ||
BC014414.1 | - | 3 | 1577 | 4548146 |