Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC441155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271675.1 | NP_001258604.1 | 2 | 1601 | ||
XM_017010889.1 | XP_016866378.1 | 2 | 1117 | |||
GenBank mRNA | AK302514.1 | - | 2 | 1557 |