Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093728.2 | NP_001087197.1 | 4 | 483 | ||
NM_001261817.1 | NP_001248746.1 | 4 | 468 | |||
NM_001261818.1 | NP_001248747.1 | 3 | 345 | |||
NM_001261819.1 | NP_001248748.1 | 3 | 330 | |||
NM_001261820.1 | NP_001248749.1 | 5 | 689 | |||
NM_001261821.1 | NP_001248750.1 | 5 | 674 | |||
NM_018679.5 | NP_061149.1 | 4 | 577 | |||
XM_005249339.2 | XP_005249396.1 | 3 | 238 | |||
XM_011514830.1 | XP_011513132.1 | 4 | 715 | |||
GenBank mRNA | AF260330.1 | - | 1 | 26 | ||
AF269223.1 | - | 3 | 261 | |||
AF536532.1 | - | 5 | 529 | |||
AF536533.1 | - | 4 | 516 | |||
AK057234.1 | - | 4 | 300 | |||
AK301778.1 | - | 4 | 359 | |||
AK301975.1 | - | 3 | 266 | |||
AK302022.1 | - | 3 | 231 | |||
AK302375.1 | - | 4 | 408 | |||
AK302456.1 | - | 4 | 389 | |||
AK315078.1 | - | 4 | 510 | |||
AK316403.1 | - | 4 | 292 | |||
AY069943.1 | - | 3 | 285 | |||
BC033729.1 | - | 4 | 429 | 5167179 | ||
BC040003.1 | - | 5 | 531 | 5741923 | ||
BC048418.1 | - | 4 | 507 | 5298037 | ||
BC090050.1 | - | 5 | 535 | 30387768 | ||
DC395417.1 | - | 4 | 416 |