Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGAPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020343.3 | NP_065076.2 | 32 | 4768 | ||
XM_005260768.2 | XP_005260825.1 | 32 | 4783 | |||
XM_006723598.1 | XP_006723661.1 | 33 | 4924 | |||
XM_006723599.1 | XP_006723662.1 | 33 | 4924 | |||
XM_011529309.1 | XP_011527611.1 | 33 | 4924 | |||
XM_011529310.2 | XP_011527612.1 | 31 | 4545 | |||
XM_011529311.2 | XP_011527613.2 | 19 | 4884 | |||
XM_017027972.1 | XP_016883461.1 | 33 | 4924 | |||
XM_017027973.1 | XP_016883462.1 | 30 | 4387 | |||
XM_017027974.1 | XP_016883463.1 | 30 | 4382 | |||
XM_017027975.1 | XP_016883464.1 | 32 | 4716 | |||
XM_017027976.1 | XP_016883465.1 | 30 | 4569 | |||
GenBank mRNA | AK002211.1 | - | 24 | 3637 | ||
AK127228.1 | - | 5 | 2939 | |||
BC171842.1 | - | 30 | 4569 | |||
DQ310704.1 | - | 32 | 4625 |