Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC45A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286646.1 | NP_001273575.1 | 1 | 435 | ||
XM_011517169.2 | XP_011515471.1 | 2 | 476 | |||
XM_011517170.2 | XP_011515472.1 | 3 | 1281 | |||
XM_011517171.2 | XP_011515473.1 | 2 | 531 | |||
XM_011517172.2 | XP_011515474.1 | 2 | 1020 | |||
XM_011517173.2 | XP_011515475.1 | 2 | 476 | |||
XM_011517176.2 | XP_011515478.1 | 2 | 476 | |||
XM_017013648.1 | XP_016869137.1 | 2 | 1555 | |||
XM_017013649.1 | XP_016869138.1 | 2 | 460 |