Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012293.2 | NP_036425.1 | 23 | 4551 | ||
XM_005264707.3 | XP_005264764.1 | 22 | 4467 | |||
XM_011510396.1 | XP_011508698.1 | 23 | 4308 | |||
XM_011510397.2 | XP_011508699.1 | 22 | 4396 | |||
GenBank mRNA | AF200348.1 | - | 23 | 4510 | ||
AK098167.1 | - | 2 | 1797 | |||
BC041027.1 | - | 5 | 676 | |||
BC070119.1 | - | 15 | 3546 | |||
D86983.1 | - | 23 | 4510 |