Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTTNBP2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018704.2 | NP_061174.1 | 6 | 2368 | ||
XM_011541781.2 | XP_011540083.1 | 6 | 2439 | |||
XM_017001806.1 | XP_016857295.1 | 6 | 2806 | |||
GenBank mRNA | AK022544.1 | - | 5 | 2289 | ||
BC016029.1 | - | 6 | 2308 | 3921313 |