Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033273.2 | NP_150376.1 | 5 | 1949 | ||
XM_011515604.2 | XP_011513906.1 | 6 | 2135 | |||
XM_011515608.2 | XP_011513910.1 | 6 | 2039 | |||
XM_011515609.2 | XP_011513911.1 | 4 | 1930 | |||
XM_011515610.2 | XP_011513912.1 | 7 | 2216 | |||
XM_011515611.2 | XP_011513913.1 | 5 | 2083 | |||
XM_017012777.1 | XP_016868266.1 | 4 | 1784 | |||
XM_017012778.1 | XP_016868267.1 | 5 | 1987 | |||
GenBank mRNA | AF277624.1 | - | 5 | 1949 |
Gene Symbol: ZNF733P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003952.1 | 3 | 1648 |