Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP6NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080491.3 | NP_001073960.1 | 12 | 981 | ||
NM_014688.3 | NP_055503.1 | 13 | 1312 | |||
XM_006717542.3 | XP_006717605.1 | 14 | 1138 | |||
XM_011519762.2 | XP_011518064.1 | 15 | 1471 | |||
XM_011519763.2 | XP_011518065.1 | 8 | 1472 | |||
XM_017016971.1 | XP_016872460.1 | 14 | 1145 | |||
XM_017016972.1 | XP_016872461.1 | 14 | 1283 | |||
XM_017016973.1 | XP_016872462.1 | 12 | 4308 | |||
GenBank mRNA | AB449916.1 | - | 11 | 882 | ||
AK292944.1 | - | 13 | 1102 | |||
BC042943.2 | - | 13 | 1150 | 5296060 | ||
D13644.1 | - | 13 | 1161 |