Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP1R9A SIRNA Assays ›
Gene Symbol:
PPP1R9A
Gene Name:
protein phosphatase 1 regulatory subunit 9A
Gene Aliases:
NRB1, NRBI, Neurabin-I
Chromosome Location:
Chr.7: 94907202 - 95296415 on Build GRCh38
UniGene ID:
Hs.21816
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
272450
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP1R9A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166160.1
NP_001159632.1
10
2485
NM_001166161.1
NP_001159633.1
8
2353
NM_001166162.1
NP_001159634.1
8
2353
NM_001166163.1
NP_001159635.1
8
2353
NM_017650.2
NP_060120.2
9
2419
XM_011516380.1
XP_011514682.1
11
2672
XM_011516381.1
XP_011514683.1
11
2548
XM_011516382.2
XP_011514684.1
11
2561
XM_011516383.1
XP_011514685.1
11
2602
XM_011516389.1
XP_011514691.1
9
2419
XM_017012394.1
XP_016867883.1
11
2548
XM_017012395.1
XP_016867884.1
11
2548
XM_017012396.1
XP_016867885.1
9
2543
XM_017012397.1
XP_016867886.1
11
2548
XM_017012398.1
XP_016867887.1
9
2431
XM_017012399.1
XP_016867888.1
9
2419
XM_017012400.1
XP_016867889.1
9
2419
XM_017012401.1
XP_016867890.1
10
2485
XM_017012402.1
XP_016867891.1
9
2419
XM_017012403.1
XP_016867892.1
9
2419
XM_017012404.1
XP_016867893.1
11
2548
XM_017012405.1
XP_016867894.1
9
2543
XM_017012406.1
XP_016867895.1
9
2474
XM_017012407.1
XP_016867896.1
9
2419
XM_017012408.1
XP_016867897.1
9
763
XM_017012410.1
XP_016867899.1
10
2551
XM_017012411.1
XP_016867900.1
4
5165
GenBank mRNA
AB033048.1
-
8
1071
AK096514.1
-
9
2382
BC130449.1
-
8
2168
40146480
BC144108.1
-
8
2169
BC144109.1
-
8
2168
9052629
BC150636.1
-
9
2433
9057006
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
actin filament organization
aging
calcium-mediated signaling
regulation of actin filament polymerization
neuron projection development
positive regulation of protein kinase activity
regulation of filopodium assembly
negative regulation of stress fiber assembly
regulation of synapse structural plasticity
regulation of synapse assembly
excitatory postsynaptic potential
positive regulation of dendritic spine development
regulation of dendritic spine morphogenesis
cellular response to toxic substance
negative regulation of long-term synaptic potentiation
positive regulation of long term synaptic depression
negative regulation of spontaneous neurotransmitter secretion
protein binding
protein C-terminus binding
protein phosphatase 1 binding
protein kinase binding
protein domain specific binding
protein homodimerization activity
ion channel binding
actin filament binding
GTPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline