Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX11L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_051986.1 | 3 | 836 | |||
GenBank mRNA | AM992874.1 | - | 3 | 836 |
Gene Symbol: DDX11L16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110561.1 | 3 | 836 | |||
GenBank mRNA | AM992882.1 | - | 3 | 836 |